The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271676.1 | NP_001258605.1 | 3 - 3 | 1392 | 126 |
NM_001271677.1 | NP_001258606.1 | 3 - 3 | 1241 | 126 | |
NM_008330.2 | NP_032356.1 | 2 - 2 | 1182 | 126 | |
GenBank mRNA | AK156785.1 | - | 2 - 2 | 1161 | 126 |
M63630.1 | - | 2 - 2 | 1144 | 126 |