Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Srcin1
Entrez Gene ID
56013
Gene Name
SRC kinase signaling inhibitor 1
Gene Aliases
P140, SNIP, mKIAA1684, p140Cap
Chromosome Location
Chr.11: 97509339 - 97575126 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018873.2 NP_061361.2 20 - 20 6890 137
GenBank mRNA AK051643.1 - 1 - 1 1618 137
BC048145.1 - 1 - 1 831 137

View Full Product Details