The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009265.3 | NP_033291.1 | 2 - 2 | 577 | 135 |
GenBank mRNA | AK003398.1 | - | 2 - 2 | 578 | 135 |
AK009752.1 | - | 2 - 2 | 578 | 135 | |
X91825.1 | - | 2 - 2 | 561 | 135 |