Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn2r58
Entrez Gene ID
628422
Gene Name
vomeronasal 2, receptor 58
Gene Aliases
EG628422
Chromosome Location
Chr.7: 41836881 - 41872670 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105055.1 NP_001098525.1 6 - 6 1842 126

View Full Product Details