Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Olfr49
Entrez Gene ID
18348
Gene Name
olfactory receptor 49
Gene Aliases
IC6, MOR1181
Chromosome Location
Chr.14: 54281896 - 54282925 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010991.2 NP_035121.1 1 - 1 107 134
GenBank mRNA BC145891.1 - 1 - 1 107 134
BC145893.1 - 1 - 1 107 134

View Full Product Details