Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Dnm3os
- Entrez Gene ID
-
474332
- Gene Name
- dynamin 3, opposite strand
- Gene Aliases
-
6030416H16Rik, Hag2, Npn1, P/L01
- Chromosome Location
-
Chr.1: 162217623 - 162225550 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002870.2 |
|
1 - 1 |
7850 |
115 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 199a-2 |
Mir199a-2 |
Mirn199a-2, mir-199a-2 |
|
Sense |
Chr. - 162217814 - 162217923 on Build GRCm38 |
Downstream |
methyltransferase like 13 |
Mettl13 |
5630401D24Rik |
308.1kb
|
Anti-sense |
Chr. - 162533671 - 162548345 on Build GRCm38 |
View Full Product Details