Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Skint4
Entrez Gene ID
320640
Gene Name
selection and upkeep of intraepithelial T cells 4
Gene Aliases
9530098N22Rik
Chromosome Location
Chr.4: 112072016 - 112168076 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178786.4 NP_848901.2 4 - 5 956 161
GenBank mRNA AK028795.1 - 4 - 5 957 161
AK035748.1 - 4 - 5 874 161
AK162813.1 - 5 - 6 971 161
EF494897.1 - 4 - 5 755 161
EF494898.1 - 4 - 5 755 161

View Full Product Details