Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Skint4
Entrez Gene ID
320640
Gene Name
selection and upkeep of intraepithelial T cells 4
Gene Aliases
9530098N22Rik
Chromosome Location
Chr.4: 112072016 - 112168076 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178786.4 NP_848901.2 9 - 10 1480 106
GenBank mRNA AK028550.1 - 9 - 10 1532 106
AK028795.1 - 9 - 10 1481 106
AK035748.1 - 9 - 10 1398 106
AK162813.1 - 10 - 11 1495 106
EF494897.1 - 9 - 10 1279 106

View Full Product Details