Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Unc93a
- Entrez Gene ID
-
381058
- Gene Name
- unc-93 homolog A (C. elegans)
- Gene Aliases
-
Unc93l, unc-93A
- Chromosome Location
-
Chr.17: 13108617 - 13143625 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199252.2 |
NP_954860.1 |
6 - 7 |
919 |
103 |
GenBank mRNA |
AJ422143.1 |
- |
6 - 7 |
919 |
103 |
- Gene Symbol
- Gm9992
- Entrez Gene ID
-
667055
- Gene Name
- predicted gene 9992
- Gene Aliases
-
ENSMUSG00000056133
- Chromosome Location
-
Chr.17: 7363646 - 7397328 on Build GRCm38
View Full Product Details