The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031247.3 | NP_112537.2 | 5 - 5 | 1363 | 129 |
GenBank mRNA | AB164418.1 | - | 5 - 5 | 1363 | 129 |
AF337052.1 | - | 5 - 5 | 1363 | 129 | |
BC094914.1 | - | 5 - 5 | 1343 | 129 |