The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027599.3 | NP_081875.1 | 2 - 2 | 2649 | 119 |
GenBank mRNA | AK029548.1 | - | 2 - 2 | 2651 | 119 |
AV255931.2 | - | 1 - 1 | 871 | 119 | |
BC139095.1 | - | 2 - 2 | 2274 | 119 | |
BC139096.1 | - | 2 - 2 | 2274 | 119 |