Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbtb25
Entrez Gene ID
109929
Gene Name
zinc finger and BTB domain containing 25
Gene Aliases
2810462M08Rik, 2900064P18Rik, AI842128, Kup, Zfp-50, Zfp50
Chromosome Location
Chr.12: 76348900 - 76370218 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172104.1 NP_001165575.1 3 - 3 1263 68
NM_028356.2 NP_082632.2 3 - 3 1257 68
GenBank mRNA AK013374.1 - 1 - 1 534 68
BC068249.1 - 3 - 3 1233 68

View Full Product Details