The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025244.3 | NP_001020415.1 | 1 - 1 | 884 | 98 |
GenBank mRNA | AK037252.1 | - | 1 - 1 | 887 | 98 |