Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Txlng
Entrez Gene ID
353170
Gene Name
taxilin gamma
Gene Aliases
4932441K18Rik, Fiat, Lrp
Chromosome Location
Chr.X: 162778917 - 162829454 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290776.1 NP_001277705.1 9 - 10 1111 125
NM_178935.5 NP_849266.1 8 - 9 1258 125
GenBank mRNA AK030100.1 - 8 - 9 1205 125
AK031023.1 - 6 - 7 2797 125
AK031783.1 - 8 - 9 1172 125
AK044130.1 - 8 - 9 1207 125
BC117898.1 - 9 - 10 1052 125
BC117899.1 - 9 - 10 1052 125
DQ316984.1 - 8 - 9 1258 125
DQ320011.1 - 9 - 10 1111 125

View Full Product Details