The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290776.1 | NP_001277705.1 | 10 - 11 | 1220 | 115 |
NM_178935.5 | NP_849266.1 | 9 - 10 | 1367 | 115 | |
GenBank mRNA | AK030100.1 | - | 9 - 10 | 1314 | 115 |
AK031023.1 | - | 7 - 8 | 2906 | 115 | |
AK031783.1 | - | 9 - 10 | 1281 | 115 | |
AK044130.1 | - | 9 - 10 | 1316 | 115 | |
BC117898.1 | - | 10 - 11 | 1161 | 115 | |
BC117899.1 | - | 10 - 11 | 1161 | 115 | |
DQ316984.1 | - | 9 - 10 | 1367 | 115 | |
DQ320011.1 | - | 10 - 11 | 1220 | 115 |