Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Agap2
Entrez Gene ID
216439
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 2
Gene Aliases
AGAP-2, Centg1, PIKE, cnt-g1, mKIAA0167
Chromosome Location
Chr.10: 127075258 - 127093173 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033263.5 NP_001028435.1 3 - 4 2008 75
NM_001301014.1 NP_001287943.1 3 - 4 2008 75
GenBank mRNA AK147446.1 - 3 - 4 2009 75
AK220325.1 - 3 - 4 883 75

View Full Product Details