Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Uap1l1
Entrez Gene ID
227620
Gene Name
UDP-N-acteylglucosamine pyrophosphorylase 1-like 1
Gene Aliases
5730445F03Rik
Chromosome Location
Chr.2: 25361489 - 25365686 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033293.2 NP_001028465.1 7 - 8 1375 95
GenBank mRNA AK077550.1 - 7 - 8 1399 95
AK077912.1 - - 1374 95
AK131159.1 - 7 - 8 1276 95
AK149178.1 - 7 - 8 1377 95
AK151942.1 - 7 - 8 1376 95
AK159787.1 - 7 - 8 1386 95
AK168977.1 - 7 - 8 1379 95
BC043307.1 - - 1371 95
BC054408.1 - 1 - 2 1036 95
BC068207.1 - 5 - 6 775 95
BC130030.1 - 8 - 9 1358 95

View Full Product Details