The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013725.4 | NP_038753.1 | 4 - 5 | 435 | 82 |
GenBank mRNA | AK002987.1 | - | 1 - 2 | 77 | 82 |
AK005147.1 | - | 4 - 5 | 380 | 82 | |
AK011207.1 | - | - | 938 | 82 | |
AK150738.1 | - | 4 - 5 | 410 | 82 | |
BC012641.1 | - | 4 - 5 | 449 | 82 | |
CJ044201.1 | - | 4 - 5 | 436 | 82 | |
U93864.1 | - | 4 - 5 | 365 | 82 |