The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198674.2 | NP_941076.1 | 10 - 10 | 1774 | 94 |
GenBank mRNA | AK054298.1 | - | 10 - 10 | 1774 | 94 |