Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P2ry6
Entrez Gene ID
233571
Gene Name
pyrimidinergic receptor P2Y, G-protein coupled, 6
Gene Aliases
2010204J23Rik, P2Y6
Chromosome Location
Chr.7: 100937630 - 100975492 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183168.2 NP_898991.1 3 - 3 1836 117
GenBank mRNA AK142680.1 - 3 - 3 1861 117
AX775901.1 - 3 - 3 1796 117
BC027331.1 - 3 - 3 1832 117
BC064095.1 - 3 - 3 1816 117

View Full Product Details