The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039155.1 | NP_001034244.1 | 5 - 6 | 882 | 88 |
NM_001039156.1 | NP_001034245.1 | 5 - 6 | 882 | 88 | |
NM_138579.4 | NP_613045.3 | 5 - 6 | 882 | 88 | |
GenBank mRNA | DQ228000.1 | - | 5 - 6 | 882 | 88 |
DQ228001.2 | - | 5 - 6 | 882 | 88 | |
DQ228002.2 | - | 5 - 6 | 882 | 88 | |
DQ278602.1 | - | 4 - 5 | 501 | 88 |