Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cxcr1
Entrez Gene ID
227288
Gene Name
chemokine (C-X-C motif) receptor 1
Gene Aliases
Il8ra
Chromosome Location
Chr.1: 74191785 - 74198144 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178241.4 NP_839972.1 2 - 2 1038 68
GenBank mRNA AY227797.1 - 1 - 1 990 68
AY390263.1 - 2 - 2 1038 68
AY749637.1 - 1 - 1 1364 68

View Full Product Details