The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010565.3 | NP_034695.1 | 2 - 2 | 1149 | 146 |
GenBank mRNA | AK131946.1 | - | 2 - 2 | 749 | 146 |
BC026140.1 | - | 2 - 2 | 1047 | 146 | |
U95962.1 | - | 2 - 2 | 1149 | 146 | |
X90819.1 | - | 2 - 2 | 1132 | 146 |