Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Olfr8
- Entrez Gene ID
-
18372
- Gene Name
- olfactory receptor 8
- Gene Aliases
-
GA_x5J8B7TNKDU-1250-1574, MOR139-5P
- Chromosome Location
-
Chr.10: 78955207 - 78956139 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_207201.1 |
NP_997084.1 |
1 - 1 |
449 |
133 |
- Gene Symbol
- Olfr1354
- Entrez Gene ID
-
259163
- Gene Name
- olfactory receptor 1354
- Gene Aliases
-
EG257869, GA_x5J8B7TPDN2-1058-1265, GA_x6K02T03FR9-4826-3919, MOR139-5, MOR139-7, MOR185-8, Olfr1354-ps1, Olfr233-ps1
- Chromosome Location
-
Chr.10: 78916842 - 78917936 on Build GRCm38
- Gene Symbol
- Olfr1355
- Entrez Gene ID
-
257734
- Gene Name
- olfactory receptor 1355
- Gene Aliases
-
GA_x5J8B7W4DUT-12263-11330, MOR139-6
- Chromosome Location
-
Chr.10: 78875549 - 78880106 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_207571.2 |
NP_997454.1 |
2 - 2 |
664 |
133 |
View Full Product Details