Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mcc
Entrez Gene ID
328949
Gene Name
mutated in colorectal cancers
Gene Aliases
D18Ertd451e, E330037C19
Chromosome Location
Chr.18: 44425060 - 44812182 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085373.1 NP_001078842.1 19 - 19 7617 145
NM_001085374.1 NP_001078843.1 17 - 17 7585 145
GenBank mRNA AK162662.1 - 1 - 1 4635 145
BU756994.1 - 1 - 1 145

View Full Product Details