Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ces1b
Entrez Gene ID
382044
Gene Name
carboxylesterase 1B
Gene Aliases
Gm5158
Chromosome Location
Chr.8: 93056725 - 93080021 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081372.1 NP_001074841.1 6 - 7 849 101

View Full Product Details