The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146563.1 | NP_666774.1 | 1 - 2 | 180 | 99 |
GenBank mRNA | BC120866.1 | - | 1 - 2 | 79 | 99 |
BC120868.1 | - | 1 - 2 | 79 | 99 |