The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133800.3 | NP_598561.2 | 5 - 6 | 545 | 67 |
GenBank mRNA | AK036891.1 | - | 5 - 6 | 488 | 67 |
AK037035.1 | - | 5 - 6 | 539 | 67 | |
AK083539.1 | - | - | 547 | 67 | |
AK167446.1 | - | 5 - 6 | 539 | 67 | |
AK177972.1 | - | 4 - 5 | 324 | 67 | |
BC013701.1 | - | 5 - 6 | 530 | 67 | |
BC094424.1 | - | 5 - 6 | 532 | 67 |