Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bod1
Entrez Gene ID
69556
Gene Name
biorientation of chromosomes in cell division 1
Gene Aliases
2310022M17Rik, BOD2.1, Fam44b
Chromosome Location
Chr.11: 31665150 - 31671862 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024919.1 NP_001020090.1 2 - 3 486 76
GenBank mRNA AK009485.1 - 2 - 3 468 76
AK219559.1 - 1 - 2 137 76
BB616782.1 - 2 - 3 510 76
BC116435.1 - 3 - 4 318 76
BC116436.1 - 3 - 4 318 76
BC128508.1 - 3 - 4 388 76
BC128509.1 - 3 - 4 318 76

View Full Product Details