The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023371.3 | NP_075860.1 | 4 - 4 | 3447 | 148 |
GenBank mRNA | AK054045.1 | - | 4 - 4 | 3448 | 148 |