The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144919.2 | NP_659168.1 | 10 - 10 | 2421 | 102 |
GenBank mRNA | AK045994.1 | - | 10 - 10 | 2433 | 102 |
BC016208.1 | - | 10 - 10 | 2421 | 102 |