The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009757.4 | NP_033887.1 | 2 - 2 | 2855 | 124 |
GenBank mRNA | AK028735.1 | - | 2 - 2 | 2541 | 124 |
AK139773.1 | - | 2 - 2 | 2856 | 124 | |
AK139847.1 | - | 2 - 2 | 2857 | 124 | |
BC055363.1 | - | 2 - 2 | 2851 | 124 |