Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn2r88
Entrez Gene ID
669149
Gene Name
vomeronasal 2, receptor 88
Gene Aliases
V2r13, V2r3
Chromosome Location
Chr.14: 51411001 - 51419527 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011686.1 NP_035816.1 2 - 3 512 79

View Full Product Details