Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Zxda
- Entrez Gene ID
-
668171
- Gene Name
- zinc finger, X-linked, duplicated A
- Gene Aliases
-
4933406P09Rik, AW413531
- Chromosome Location
-
Chr.X: 94791285 - 94798289 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003292.1 |
|
1 - 1 |
3442 |
91 |
GenBank mRNA |
AK155431.1 |
- |
2 - 2 |
2686 |
91 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB653381.1 |
- |
1 |
480 |
|
Chr. X - 94794676 - 94795327 [-] on Build GRCm38 |
91 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
predicted gene, 39526 |
Gm39526 |
|
-44.4kb
|
Anti-sense |
Chr. - 94730610 - 94746915 on Build GRCm38 |
Downstream |
spindlin family, member 4 |
Spin4 |
9630042H07Rik |
224.2kb
|
Sense |
Chr. - 95022507 - 95026682 on Build GRCm38 |
View Full Product Details