Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Krt36
- Entrez Gene ID
-
16673
- Gene Name
- keratin 36
- Gene Aliases
-
HRa-1, K36, Krt1-22, Krt1-5, MHRa-1
- Chromosome Location
-
Chr.11: 100102012 - 100114634 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA725946.1 |
- |
1 |
|
|
Chr. 11 - 100102013 - 100102936 [+] on Build GRCm38 |
73 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
keratin 35 |
Krt35 |
AI662400, Ha5, Krt1-24, Krtha5 |
-5.8kb
|
Sense |
Chr. - 100092192 - 100096224 on Build GRCm38 |
Downstream |
keratin 13 |
Krt13 |
CK13, K13, Krt-1.13, Krt1-13 |
15.2kb
|
Sense |
Chr. - 100117596 - 100121495 on Build GRCm38 |
View Full Product Details