Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pramel4
Entrez Gene ID
347710
Gene Name
preferentially expressed antigen in melanoma like 4
Gene Aliases
C79430, D4Ertd792e
Chromosome Location
Chr.4: 144059126 - 144069324 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001319.3 NP_001001319.2 3 - 4 1015 107
GenBank mRNA AF490340.1 - 3 - 4 899 107
AK145585.1 - 3 - 4 1016 107
AK163305.1 - 3 - 4 874 107

Gene Symbol
Gm13102
Entrez Gene ID
100041077
Gene Name
predicted gene 13102
Gene Aliases
OTTMUSG00000010432
Chromosome Location
Chr.4: 144099880 - 144110101 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085419.1 NP_001078888.1 3 - 4 1014 107
GenBank mRNA BC132592.1 - 3 - 4 1015 107
BC132620.1 - 3 - 4 1015 107

View Full Product Details