The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136235.1 | NP_001129707.1 | 2 - 3 | 360 | 94 |
GenBank mRNA | BG088171.2 | - | 2 - 3 | 360 | 94 |