The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029872.1 | NP_084148.1 | 1 - 1 | 650 | 76 |
GenBank mRNA | AK019388.1 | - | 1 - 1 | 675 | 76 |