Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn2r98
Entrez Gene ID
224552
Gene Name
vomeronasal 2, receptor 98
Gene Aliases
EG224552
Chromosome Location
Chr.17: 19053493 - 19081311 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104550.1 NP_001098020.1 6 - 6 1952 82

View Full Product Details