Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sypl2
- Entrez Gene ID
-
17306
- Gene Name
- synaptophysin-like 2
- Gene Aliases
-
AI552439, Mg29
- Chromosome Location
-
Chr.3: 108210410 - 108226669 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK005920.1 |
- |
1 - 1 |
253 |
69 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BU939392.1 |
- |
1 |
347 |
|
Chr. 3 - 108225571 - 108226063 [-] on Build GRCm38 |
69 |
GenBankmRNA |
CA465870.1 |
- |
|
323 |
|
Chr. - - [-] on Build |
69 |
GenBankmRNA |
BY706058.1 |
- |
1 |
253 |
|
Chr. 3 - 108225572 - 108225968 [-] on Build GRCm38 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ataxin 7-like 2 |
Atxn7l2 |
2610528J18Rik |
-15.0kb
|
Sense |
Chr. - 108202222 - 108210527 on Build GRCm38 |
Downstream |
predicted gene, 40123 |
Gm40123 |
|
840.0
|
Anti-sense |
Chr. - 108226808 - 108239467 on Build GRCm38 |
View Full Product Details