Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dera
Entrez Gene ID
232449
Gene Name
2-deoxyribose-5-phosphate aldolase homolog (C. elegans)
Gene Aliases
2010002D22Rik, 2500002K03Rik, DEOC
Chromosome Location
Chr.6: 137754577 - 137837872 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK008033.1 - 1 - 2 93 101


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY708021.1 - 1 - 2 93 Chr. 6 - 137754546 - 137760963 [+] on Build GRCm38 101


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream serine/threonine kinase receptor associated protein Strap AW557906, C78091, C79202, Unrip -2.6kb Sense Chr. - 137735082 - 137751930 on Build GRCm38
Downstream solute carrier family 15, member 5 Slc15a5 9830102E05Rik 222.6kb Anti-sense Chr. - 137983590 - 138079916 on Build GRCm38

View Full Product Details