Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Atpaf1
- Entrez Gene ID
-
230649
- Gene Name
- ATP synthase mitochondrial F1 complex assembly factor 1
- Gene Aliases
-
6330547J17Rik, AI593252, ATP11, ATP11p
- Chromosome Location
-
Chr.4: 115784814 - 115812314 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK029823.1 |
- |
1 - 2 |
83 |
96 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
testis expressed 38 |
Tex38 |
4930544O15Rik, Theg4 |
-18.4kb
|
Sense |
Chr. - 115779831 - 115782759 on Build GRCm38 |
Downstream |
MOB kinase activator 3C |
Mob3c |
AW822253, D130076I06Rik, Mobkl2c |
7.3kb
|
Anti-sense |
Chr. - 115818756 - 115836185 on Build GRCm38 |
View Full Product Details