Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- A330087I24
- Entrez Gene ID
-
327857
- Gene Name
- uncharacterized protein A330087I24
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK039689.1 |
- |
1 - 2 |
297 |
65 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
vomeronasal 2, receptor 87 |
Vmn2r87 |
EG625131 |
|
Sense |
Chr. - 130471821 - 130497379 on Build GRCm38 |
View Full Product Details