Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- 4933431E20Rik
- Entrez Gene ID
-
329735
- Gene Name
- RIKEN cDNA 4933431E20 gene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 107888850 - 107896213 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_015459.1 |
|
3 - 3 |
2867 |
66 |
GenBank mRNA |
AK045063.1 |
- |
1 - 1 |
52 |
66 |
AK147824.1 |
- |
4 - 4 |
2464 |
66 |
BC060136.1 |
- |
3 - 3 |
2867 |
66 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB643293.1 |
- |
1 |
49 |
|
Chr. 3 - 107890771 - 107891438 [-] on Build GRCm38 |
66 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
colony stimulating factor 1 (macrophage) |
Csf1 |
C87615, Csfm, MCSF, op |
-129.4kb
|
Sense |
Chr. - 107741048 - 107760469 on Build GRCm38 |
Downstream |
glutathione S-transferase, mu 5 |
Gstm5 |
|
4.4kb
|
Anti-sense |
Chr. - 107895854 - 107898685 on Build GRCm38 |
View Full Product Details