Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sdc2
- Entrez Gene ID
-
15529
- Gene Name
- syndecan 2
- Gene Aliases
-
4833414L08Rik, AA960457, Hspg1, Synd2, syndecan-2
- Chromosome Location
-
Chr.15: 32920723 - 33034721 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK047924.1 |
- |
1 - 1 |
2255 |
97 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
predicted gene, 32618 |
Gm32618 |
|
-228.5kb
|
Sense |
Chr. - 32778650 - 32795299 on Build GRCm38 |
Downstream |
carboxypeptidase Q |
Cpq |
1190003P12Rik, 2610034C17Rik, Hls2, Lal-1, Pgcp |
56.7kb
|
Sense |
Chr. - 33083129 - 33594552 on Build GRCm38 |
View Full Product Details