Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bnc2
Entrez Gene ID
242509
Gene Name
basonuclin 2
Gene Aliases
5031434M05Rik, 8430420F16Rik
Chromosome Location
Chr.4: 84266804 - 84676342 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172870.4 NP_766458.3 6 - 6 5512 68
GenBank mRNA AK018429.1 - 1 - 1 450 68


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CA983934.1 - 1 422 Chr. 4 - 84273055 - 84273904 [-] on Build GRCm38 68
GenBankmRNA BB620417.1 - 1 448 Chr. 4 - 84273282 - 84273931 [-] on Build GRCm38 68
GenBankmRNA CN533598.1 - 1 437 Chr. 4 - 84273248 - 84273919 [-] on Build GRCm38 68


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream coiled-coil domain containing 171 Ccdc171 4930418J05Rik, 4930473A06Rik, A330015D16Rik -407.1kb Anti-sense Chr. - 83525537 - 83866143 on Build GRCm38
Downstream centlein, centrosomal protein Cntln AI448092, B430108F07Rik, D530005L17Rik 609.6kb Anti-sense Chr. - 84883512 - 85131923 on Build GRCm38

View Full Product Details