Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rem1
- Entrez Gene ID
-
19700
- Gene Name
- rad and gem related GTP binding protein 1
- Gene Aliases
-
E030011C07Rik, Ras, Rem
- Chromosome Location
-
Chr.2: 152627008 - 152635191 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK033315.1 |
- |
- |
3050 |
79 |
AK078772.1 |
- |
- |
3051 |
79 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB068330.2 |
- |
1 |
540 |
|
Chr. 2 - 152637351 - 152637996 [+] on Build GRCm38 |
79 |
GenBankmRNA |
BB070632.2 |
- |
1 |
524 |
|
Chr. 2 - 152637346 - 152637996 [+] on Build GRCm38 |
79 |
GenBankmRNA |
BB071316.1 |
- |
1 |
158 |
|
Chr. 2 - 152637710 - 152637996 [+] on Build GRCm38 |
79 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Downstream |
RIKEN cDNA 1700030C14 gene |
1700030C14Rik |
|
14.9kb
|
Anti-sense |
Chr. - 152652882 - 152654348 on Build GRCm38 |
View Full Product Details