Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- A230005M16Rik
- Entrez Gene ID
-
654807
- Gene Name
- RIKEN cDNA A230005M16 gene
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK082064.1 |
- |
1 - 2 |
99 |
72 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB649961.1 |
- |
1 - 2 |
95 |
|
Chr. 2 - 25707245 - 25709712 [-] on Build GRCm38 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
odorant binding protein 2A |
Obp2a |
BC027556, Lcn13 |
-2.5kb
|
Anti-sense |
Chr. - 25697543 - 25703326 on Build GRCm38 |
Downstream |
odorant binding protein 2B |
Obp2b |
Lcn14, OTTMUSG00000012631 |
4.4kb
|
Anti-sense |
Chr. - 25714105 - 25740115 on Build GRCm38 |
View Full Product Details