Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tbp
- Entrez Gene ID
-
21374
- Gene Name
- TATA box binding protein
- Gene Aliases
-
GTF2D1, Gtf2d, SCA17, TFIID
- Chromosome Location
-
Chr.17: 15499888 - 15517427 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK089397.1 |
- |
1 - 1 |
1144 |
72 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG078278.2 |
- |
1 |
399 |
|
Chr. 17 - 15500663 - 15501237 [+] on Build GRCm38 |
72 |
GenBankmRNA |
BY764398.1 |
- |
1 |
295 |
|
Chr. 17 - 15500768 - 15501482 [+] on Build GRCm38 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
proteasome (prosome, macropain) subunit, beta type 1 |
Psmb1 |
AA409053, C81484, Lmpc5 |
-1.6kb
|
Anti-sense |
Chr. - 15475721 - 15498276 on Build GRCm38 |
Downstream |
programmed cell death 2 |
Pdcd2 |
AI528543, Pcd2, RP-8 |
20.1kb
|
Anti-sense |
Chr. - 15521575 - 15527301 on Build GRCm38 |
View Full Product Details