Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Wdfy2
- Entrez Gene ID
-
268752
- Gene Name
- WD repeat and FYVE domain containing 2
- Gene Aliases
-
5830485M08, B130024L21Rik, ProF, ZFYVE22
- Chromosome Location
-
Chr.14: 62837690 - 62961509 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK158235.1 |
- |
- |
86 |
57 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY753019.1 |
- |
1 - 2 |
83 |
|
Chr. 14 - 62837677 - 62838205 [-] on Build GRCm38 |
57 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 8098 |
Mir8098 |
Gm27462, mmu-mir-8098 |
|
Anti-sense |
Chr. - 62837721 - 62837857 on Build GRCm38 |
Downstream |
microRNA 6541 |
Mir6541 |
Gm27311, mmu-mir-6541 |
27.3kb
|
Anti-sense |
Chr. - 62865542 - 62865651 on Build GRCm38 |
View Full Product Details